missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonamide, 98%

Catalog No. AAL0908203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0908203 1 g
AAL0908206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0908203 Supplier Thermo Scientific Chemicals Supplier No. L0908203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6961-82-6 | C6H6ClNO2S | 191.629 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6961-82-6
Molecular Formula C6H6ClNO2S
Molecular Weight (g/mol) 191.629
MDL Number MFCD00051974
InChI Key JCCBZCMSYUSCFM-UHFFFAOYSA-N
Synonym o-chlorobenzenesulfonamide, 2-chlorobenzene-1-sulfonamide, benzenesulfonamide, 2-chloro, unii-4bt2i28a0c, benzenesulfonamide, o-chloro, 2-chlorobenzenesulphonamide, 2-chloro-benzenesulfonamide, chlorobenzenesulfonamide, o-chlorobenzenesulphonamide, benzenesulfonamide, chloro
PubChem CID 81410
IUPAC Name 2-chlorobenzenesulfonamide
SMILES C1=CC=C(C(=C1)S(=O)(=O)N)Cl

Specifications

Melting Point 189°C to 191°C
Quantity 1 g
Beilstein 2937540
Formula Weight 191.64
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzenesulfonamide
EINECSNumber 230-156-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only