missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzaldehyde 99.0+%, TCI America™

Catalog No. C056125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C056125G 25 g
C0561500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C056125G Supplier TCI America Supplier No. C056125G

Chemical Identifiers

CAS 89-98-5
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.566
MDL Number MFCD00003304
InChI Key FPYUJUBAXZAQNL-UHFFFAOYSA-N
Synonym o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
PubChem CID 6996
IUPAC Name 2-chlorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)Cl

Specifications

Color Yellow
Boiling Point 212°C
Quantity 25 g
UN Number 3265
Formula Weight 140.57
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzaldehyde
EINECSNumber (3)-1162
RTECSNumber CU5075000
TSCA Yes