missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzaldehyde, 97%

CAS: 89-98-5 | C7H5ClO | 140.566 g/mol

Supplier:  Thermo Scientific Chemicals A134880B

 View more versions of this product

Catalog No. AAA134880B


Only null left
Explore available promotions

Description

Description

2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzaldehyde has been used in generation of small focused library of diversely functionalized dihydropyrimidine derivatives via one-pot three-component Biginelli cyclocondensation of β-ketoesters, aldehydes and thioureas

Solubility
Soluble in water (1.8g/L), alcohol, ether, benzene, and acetone.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Chlorobenzaldehyde
10°C to 12°C
2.9
87°C (188°F)
C7H5ClO
MFCD00003304
UN3265
Air Sensitive
Soluble in water (1.8g/L),alcohol,ether,benzene,and acetone.
C1=CC=C(C(=C1)C=O)Cl
140.566
140.57
89-98-5
1.249
212°C
Pungent
1.5675
1000 g
385877
o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
FPYUJUBAXZAQNL-UHFFFAOYSA-N
2-chlorobenzaldehyde
6996
97%

Chemical Identifiers

89-98-5
140.566
FPYUJUBAXZAQNL-UHFFFAOYSA-N
6996
C1=CC=C(C(=C1)C=O)Cl
C7H5ClO
MFCD00003304
o-chlorobenzaldehyde, chlorobenzaldehyde, benzaldehyde, 2-chloro, benzaldehyde, o-chloro, 2-chlorbenzaldehyd, 2-clorobenzaldeide, o-chloorbenzaldehyde, usaf m-7, 2-chloorbenzaldehyde, o-chlorobenzenecarboxaldehyde
2-chlorobenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 201-956-3

RTECSNumber : CU5075000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only