missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroanthracene 98.0+%, TCI America™

Catalog No. C19621G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19621G Supplier TCI America Supplier No. C19621G
Only null left

Chemical Identifiers

CAS 17135-78-3
Molecular Formula C14H9Cl
Molecular Weight (g/mol) 212.676
InChI Key OWFINXQLBMJDJQ-UHFFFAOYSA-N
Synonym anthracene, 2-chloro, ccris 5549, .beta.-chloro anthracene, 2-chloroanthracene, acmc-1c5s8, dsstox_cid_29086, dsstox_rid_83305, dsstox_gsid_49230, 4-05-00-02292 beilstein handbook reference, owfinxqlbmjdjq-uhfffaoysa
PubChem CID 28308
IUPAC Name 2-chloroanthracene
SMILES C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Cl

Specifications

Melting Point 224°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroanthracene
RTECSNumber CA9560000
TSCA No