accessibility menu, dialog, popup

UserName

Cl

CAS RN 17135-78-3

IUPAC Name: 2-chloroanthracene
Synonyms: 2-chloroanthracene
Molecular Weight (g/mol): 212.68
Molecular Formula: C14H9Cl
InChi Key: OWFINXQLBMJDJQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C=C3C=CC=CC3=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results