missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroanisole, 98%

Catalog No. AAL0695218
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0695218 50 g
AAL0695230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0695218 Supplier Thermo Scientific Chemicals Supplier No. L0695218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 766-51-8 | C7H7ClO | 142.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-51-8
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00000557
InChI Key QGRPVMLBTFGQDQ-UHFFFAOYSA-N
Synonym 2-chloroanisole, o-chloroanisole, benzene, 1-chloro-2-methoxy, anisole, o-chloro, o-chloromethoxybenzene, o-chlorophenyl methyl ether, 2-chlorophenol methyl ether, chloroanisole, 1-chloro-2-methoxy-benzene, labotest-bb ltbb002921
PubChem CID 13011
IUPAC Name 1-chloro-2-methoxybenzene
SMILES COC1=CC=CC=C1Cl

Specifications

Density 1.123
Boiling Point 194°C to 196°C
Flash Point 76°C (168°F)
Refractive Index 1.545
Quantity 50 g
Beilstein 2042179
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 2-Chloroanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 212-167-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only