accessibility menu, dialog, popup

UserName

CAS RN 766-51-8

IUPAC Name: 1-chloro-2-methoxybenzene
Synonyms: O-chloroanisole
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: QGRPVMLBTFGQDQ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1Cl
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)