missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals

Catalog No. p-7053442
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5994803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5994803 Supplier Thermo Scientific Chemicals Supplier No. H5994803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6526-67-6 | Molecular Formula: C9H10ClNO | Molecular Weight: 183.64 g/mol

2-Chloro-N,N-dimethylbenzamide is involved in the preparation of repellents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N,N-dimethylbenzamide is involved in the preparation of repellents.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6526-67-6
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.64
MDL Number MFCD00463636
InChI Key SZUQPOLAEFBJJH-UHFFFAOYSA-N
Synonym benzamide, 2-chloro-n,n-dimethyl, n,n-dimethyl-2-chlorobenzamide, o-chloro-n,n-dimethylbenzamide, 2-chloro-n,n-dimethyl-benzamide, benzamide, o-chloro-n,n-dimethyl, aronis25704, 2-chlorophenyl-n,n-dimethylcarboxamide
PubChem CID 81012
IUPAC Name 2-chloro-N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 183.64
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chloro-N,N-dimethylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only