accessibility menu, dialog, popup

UserName

CH3NH3COCl

CAS RN 6526-67-6

IUPAC Name: 2-chloro-N,N-dimethylbenzamide
Synonyms: 2-chloro-N,N-dimethylbenzamide
Molecular Weight (g/mol): 183.64
Molecular Formula: C9H10ClNO
InChi Key: SZUQPOLAEFBJJH-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results