missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(2,3-dimethylphenyl)benzamide, 97%

Catalog No. AAH56225MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56225MD 250 mg
AAH5622503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56225MD Supplier Thermo Scientific Chemicals Supplier No. H56225MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 196617-88-6 | C15H14ClNO | 259.733 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 196617-88-6
Molecular Formula C15H14ClNO
Molecular Weight (g/mol) 259.733
MDL Number MFCD00018254
InChI Key OUAGJVUPQKYHDU-UHFFFAOYSA-N
Synonym 2-chloro-n-2,3-dimethylphenyl benzamide, cambridge id 5328590, 2-chloro-2',3'-benzoxylidide, 2-chloro-n-2,3-dimethyl-phenyl-benzamide, n-2,3-dimethylphenyl 2-chlorophenyl carboxamide
PubChem CID 295079
IUPAC Name 2-chloro-N-(2,3-dimethylphenyl)benzamide
SMILES CC1=C(C(=CC=C1)NC(=O)C2=CC=CC=C2Cl)C

Specifications

Quantity 250 mg
Formula Weight 259.74
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-(2,3-dimethylphenyl)benzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only