accessibility menu, dialog, popup

UserName

CAS RN 196617-88-6

IUPAC Name: 2-chloro-N-(2,3-dimethylphenyl)benzamide
Synonyms: 2-chloro-N-(2,3-dimethylphenyl)benzamide
Molecular Weight (g/mol): 259.73
Molecular Formula: C15H14ClNO
InChi Key: OUAGJVUPQKYHDU-UHFFFAOYSA-N
SMILES: CC1=C(C)C(NC(=O)C2=CC=CC=C2Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)