missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrophenol 98.0+%, TCI America™

Catalog No. C30745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30745G 5 g
C30741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30745G Supplier TCI America Supplier No. C30745G

Chemical Identifiers

CAS 603-86-1
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD00024233
InChI Key ICCYFVWQNFMENX-UHFFFAOYSA-N
Synonym 2-chloro-6-nitrophenol, phenol, 2-chloro-6-nitro, 6-chloro-2-nitrophenol, 2-chloro-6-nitro-phenol, pubchem4103, 2-nitro-6-chlorophenol, acmc-1aych, 2-chloro-6-nitro-pheno, 2-chlor-6-nitro-phenol, ksc494c1j
PubChem CID 11784
IUPAC Name 2-chloro-6-nitrophenol
SMILES OC1=C(Cl)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 71°C
Color Yellow
Quantity 5 g
Formula Weight 173.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-nitrophenol