accessibility menu, dialog, popup

UserName

OHClNOO

CAS RN 603-86-1

IUPAC Name: 2-chloro-6-nitrophenol
Synonyms: 6-chloro-2-nitrophenol
Molecular Weight (g/mol): 173.55
Molecular Formula: C6H4ClNO3
InChi Key: ICCYFVWQNFMENX-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)