missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrophenol, 98%

Catalog No. AAA1717203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1717203 1 g
AAA1717206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1717203 Supplier Thermo Scientific Chemicals Supplier No. A1717203

CAS: 603-86-1 | C6H4ClNO3 | 173.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 603-86-1
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.55
MDL Number MFCD00024233
InChI Key ICCYFVWQNFMENX-UHFFFAOYSA-N
Synonym 2-chloro-6-nitrophenol, phenol, 2-chloro-6-nitro, 6-chloro-2-nitrophenol, 2-chloro-6-nitro-phenol, pubchem4103, 2-nitro-6-chlorophenol, acmc-1aych, 2-chloro-6-nitro-pheno, 2-chlor-6-nitro-phenol, ksc494c1j
PubChem CID 11784
IUPAC Name 2-chloro-6-nitrophenol
SMILES OC1=C(Cl)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 69°C to 71°C
Odor Characteristic
Quantity 1 g
Formula Weight 173.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-nitrophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-061-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only