missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-nitrobenzonitrile, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 118°C to 124°C |
| Molecular Formula | C8H6ClN |
| Quantity | 5 g |
| Beilstein | 1369781 |
| Synonym | benzonitrile, 2-chloro-6-methyl, 2-cyano-3-chlorotoluene, 3-chloro-2-toluonitrile, o-tolunitrile, 6-chloro, 6-chloro-o-tolunitrile, unii-41m65jiw54, 2-chloro-6-methyl benzonitrile, 2-chloro-6-methylbenzenecarbonitrile, 2-cyano3-chlorotoluene, pubchem23832 |
| InChI Key | WQWQHJNUHQEGTN-UHFFFAOYSA-N |
| SMILES | CC1=C(C(=CC=C1)Cl)C#N |
| IUPAC Name | 2-chloro-6-methylbenzonitrile |
| Molecular Weight (g/mol) | 151.59 |
| PubChem CID | 23053 |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 229-496-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only