accessibility menu, dialog, popup

UserName

ONOClN

CAS RN 6575-07-1

IUPAC Name: 2-chloro-6-nitrobenzonitrile
Synonyms: 2-chloro-6-nitrobenzonitrile
Molecular Weight (g/mol): 182.56
Molecular Formula: C7H3ClN2O2
InChi Key: ZHLCARBDIRRRHD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(Cl)=C1C#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results