missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzonitrile, 98%

Catalog No. AAA1270314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1270314 25 g
AAA1270306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1270314 Supplier Thermo Scientific Chemicals Supplier No. A1270314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6575-07-1 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6575-07-1
MDL Number MFCD00007203

Specifications

Melting Point 118°C to 124°C
Molecular Formula C7H3ClN2O2
Quantity 25 g
Beilstein 1369781
Synonym 6-chloro-2-nitrobenzonitrile, 2-chloro-6-nitro-benzonitrile, 3-chloro-2-cyanonitrobenzene, benzonitrile, 2-chloro-6-nitro, 6-chloro-2-nitrobenzenecarbonitrile, acmc-20anya, pubchem4742, 7-tert-butylisophthalic acid, 2-nitro-6-chlorobenzonitrile, 2-nitro-6-chloro-benzonitrile
InChI Key ZHLCARBDIRRRHD-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)C#N)[N+](=O)[O-]
IUPAC Name 2-chloro-6-nitrobenzonitrile
Molecular Weight (g/mol) 182.56
PubChem CID 81053
Formula Weight 182.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-nitrobenzonitrile
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 229-496-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only