missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%

CAS: 57729-79-0 | C7H4ClF3N2O2 | 240.566 g/mol

$35.84 - $120.04

Chemical Identifiers

CAS 57729-79-0
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.566
MDL Number MFCD00042153
InChI Key JLWRJMVXRUKFPA-UHFFFAOYSA-N
Synonym 2-chloro-6-nitro-4-trifluoromethyl aniline, 4-amino-3-chloro-5-nitrobenzotrifluoride, 6-chloro-2-nitro-4-trifluoromethyl phenylamine, pubchem2811, 2-chloro-4-trifluoromethyl-6-nitroaniline, 4-amino-3-chloro-5-nitro-benzotrifluoride, 2-chloranyl-6-nitro-4-trifluoromethyl aniline, 2-chloro-6-nitro-4-trifluoromethyl-phenylamine, 2-chloro-6-nitro-4-trifluoromethyl phenylamine
PubChem CID 2734081
IUPAC Name 2-chloro-6-nitro-4-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2093803
View Documents
Thermo Scientific Chemicals
B2093803
1 g N/A
Only null left
 
AAB2093806
View Documents
Thermo Scientific Chemicals
B2093806
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57729-79-0
240.566
JLWRJMVXRUKFPA-UHFFFAOYSA-N
2734081
C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
C7H4ClF3N2O2
MFCD00042153
2-chloro-6-nitro-4-trifluoromethyl aniline, 4-amino-3-chloro-5-nitrobenzotrifluoride, 6-chloro-2-nitro-4-trifluoromethyl phenylamine, pubchem2811, 2-chloro-4-trifluoromethyl-6-nitroaniline, 4-amino-3-chloro-5-nitro-benzotrifluoride, 2-chloranyl-6-nitro-4-trifluoromethyl aniline, 2-chloro-6-nitro-4-trifluoromethyl-phenylamine, 2-chloro-6-nitro-4-trifluoromethyl phenylamine
2-chloro-6-nitro-4-(trifluoromethyl)aniline

Specifications

57729-79-0
C7H4ClF3N2O2
1 g
2-chloro-6-nitro-4-trifluoromethyl aniline, 4-amino-3-chloro-5-nitrobenzotrifluoride, 6-chloro-2-nitro-4-trifluoromethyl phenylamine, pubchem2811, 2-chloro-4-trifluoromethyl-6-nitroaniline, 4-amino-3-chloro-5-nitro-benzotrifluoride, 2-chloranyl-6-nitro-4-trifluoromethyl aniline, 2-chloro-6-nitro-4-trifluoromethyl-phenylamine, 2-chloro-6-nitro-4-trifluoromethyl phenylamine
C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F
240.566
240.58
2-Chloro-6-nitro-4-(trifluoromethyl)aniline
63°C to 67°C
MFCD00042153
UN2811
JLWRJMVXRUKFPA-UHFFFAOYSA-N
2-chloro-6-nitro-4-(trifluoromethyl)aniline
2734081
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only