CAS RN 57729-79-0
IUPAC Name:
2-chloro-6-nitro-4-(trifluoromethyl)aniline
Synonyms:
2-chloro-6-nitro-4-(trifluoromethyl)aniline
Molecular Weight (g/mol):
240.57
Molecular Formula:
C7H4ClF3N2O2
InChi Key:
JLWRJMVXRUKFPA-UHFFFAOYSA-N
SMILES:
NC1=C(Cl)C=C(C=C1[N+]([O-])=O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%
2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%
CAS: 57729-79-0 Molecular Formula: C7H4ClF3N2O2 Molecular Weight (g/mol): 240.566 MDL Number: MFCD00042153 InChI Key: JLWRJMVXRUKFPA-UHFFFAOYSA-N Synonym: 2-chloro-6-nitro-4-trifluoromethyl aniline,4-amino-3-chloro-5-nitrobenzotrifluoride,6-chloro-2-nitro-4-trifluoromethyl phenylamine,pubchem2811,2-chloro-4-trifluoromethyl-6-nitroaniline,4-amino-3-chloro-5-nitro-benzotrifluoride,2-chloranyl-6-nitro-4-trifluoromethyl aniline,2-chloro-6-nitro-4-trifluoromethyl-phenylamine,2-chloro-6-nitro-4-trifluoromethyl phenylamine PubChem CID: 2734081 IUPAC Name: 2-chloro-6-nitro-4-(trifluoromethyl)aniline SMILES: C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F