missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitro-4-(trifluoromethyl)aniline, 97%

Catalog No. AAB2093803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2093803 1 g
AAB2093806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2093803 Supplier Thermo Scientific Chemicals Supplier No. B2093803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57729-79-0 | C7H4ClF3N2O2 | 240.566 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57729-79-0
Molecular Formula C7H4ClF3N2O2
Molecular Weight (g/mol) 240.566
MDL Number MFCD00042153
InChI Key JLWRJMVXRUKFPA-UHFFFAOYSA-N
Synonym 2-chloro-6-nitro-4-trifluoromethyl aniline, 4-amino-3-chloro-5-nitrobenzotrifluoride, 6-chloro-2-nitro-4-trifluoromethyl phenylamine, pubchem2811, 2-chloro-4-trifluoromethyl-6-nitroaniline, 4-amino-3-chloro-5-nitro-benzotrifluoride, 2-chloranyl-6-nitro-4-trifluoromethyl aniline, 2-chloro-6-nitro-4-trifluoromethyl-phenylamine, 2-chloro-6-nitro-4-trifluoromethyl phenylamine
PubChem CID 2734081
IUPAC Name 2-chloro-6-nitro-4-(trifluoromethyl)aniline
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])N)Cl)C(F)(F)F

Specifications

Melting Point 63°C to 67°C
Quantity 1 g
UN Number UN2811
Formula Weight 240.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-nitro-4-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only