missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylthiophenol, 96%

Catalog No. p-7045380
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1049903 1 g
AAL1049906 5 g
AAL1049914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAL1049903 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' L1049903

CAS: 18858-05-4 | C7H7ClS | 158.643 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18858-05-4
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00041422
InChI Key WNRLJMYSWRBJIG-UHFFFAOYSA-N
Synonym 2-chloro-6-methylthiophenol, methyl thiyl chlorobenzene, acmc-1br13, 2-chloro-6-methyl-thiophenol, benzenethiol,2-chloro-6-methyl, 6-chloro-2-methylbenzene-1-thiol
PubChem CID 4130239
IUPAC Name 2-chloro-6-methylbenzenethiol
SMILES CC1=C(C(=CC=C1)Cl)S

Specifications

Boiling Point 135°C to 136°C (35 mmHg)
Refractive Index 1.595
Quantity 1 g
Beilstein 1932749
Sensitivity Air Sensitive
Formula Weight 158.65
Percent Purity 96%
Chemical Name or Material 2-Chloro-6-methylthiophenol
missing translation for 'healthHazard1' H302-H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only