Learn More
2-Chloro-6-methylphenol, 98%
CAS: 87-64-9 | C7H7ClO | 142.582 g/mol
Supplier: Thermo Scientific Chemicals L0361714
Description
2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Soluble in water (24 mg/ml at 25°C).
Notes
Storage temperature: 2 - 4°C. Incompatible with Strong oxidizing agents, strong acids and bases.
Specifications
| 2-Chloro-6-methylphenol | |
| 1.201 | |
| 72°C (161°F) | |
| 1.545 | |
| 25 g | |
| 774623 | |
| Soluble in water (24mg/ml at 25°C). | |
| CC1=C(C(=CC=C1)Cl)O | |
| 142.582 | |
| 142.59 |
| 87-64-9 | |
| 75°C to 76°C (12 mmHg) | |
| C7H7ClO | |
| MFCD00020000 | |
| UN2669 | |
| 6-chloro-o-cresol, phenol, 2-chloro-6-methyl, o-cresol, 6-chloro, 2-methyl-6-chlorophenol, 2-chloro-6-methyl-phenol, 6-chloro cresol, 6-chlor-o-kresol, pubchem3669, 6-chloro-2-methylphenol, 2-chloro-6-methyl phenol | |
| YPNZJHFXFVLXSE-UHFFFAOYSA-N | |
| 2-chloro-6-methylphenol | |
| 6898 | |
| 98% |
Chemical Identifiers
| 87-64-9 | |
| 142.582 | |
| YPNZJHFXFVLXSE-UHFFFAOYSA-N | |
| 6898 | |
| CC1=C(C(=CC=C1)Cl)O |
| C7H7ClO | |
| MFCD00020000 | |
| 6-chloro-o-cresol, phenol, 2-chloro-6-methyl, o-cresol, 6-chloro, 2-methyl-6-chlorophenol, 2-chloro-6-methyl-phenol, 6-chloro cresol, 6-chlor-o-kresol, pubchem3669, 6-chloro-2-methylphenol, 2-chloro-6-methyl phenol | |
| 2-chloro-6-methylphenol |
Safety and Handling
GHS H Statement
H331-H318-H227-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 201-760-8
RTECSNumber : GO7120100
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only