missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylphenol, 98%

Catalog No. p-7043949
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0361706 5 g
AAL0361714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0361706 Supplier Thermo Scientific Chemicals Supplier No. L0361706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 87-64-9 | C7H7ClO | 142.582 g/mol

2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-methylphenol is s an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Soluble in water (24 mg/ml at 25°C).

Notes
Storage temperature: 2 - 4°C. Incompatible with Strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 87-64-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00020000
InChI Key YPNZJHFXFVLXSE-UHFFFAOYSA-N
Synonym 6-chloro-o-cresol, phenol, 2-chloro-6-methyl, o-cresol, 6-chloro, 2-methyl-6-chlorophenol, 2-chloro-6-methyl-phenol, 6-chloro cresol, 6-chlor-o-kresol, pubchem3669, 6-chloro-2-methylphenol, 2-chloro-6-methyl phenol
PubChem CID 6898
IUPAC Name 2-chloro-6-methylphenol
SMILES CC1=C(C(=CC=C1)Cl)O

Specifications

Density 1.201
Boiling Point 75°C to 76°C (12 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.545
Quantity 5 g
UN Number UN2669
Beilstein 774623
Solubility Information Soluble in water (24mg/ml at 25°C).
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylphenol
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H331-H318-H227-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 201-760-8
RTECSNumber GO7120100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only