missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylbenzoic Acid 98.0+%, TCI America™
Supplier: TCI America C27765G
Specifications
| 2-Chloro-6-methylbenzoic Acid | |
| 106°C | |
| MFCD00045799 | |
| 6-chloro-o-toluic acid, 2-chloro-6-methyl-benzoic acid, o-toluic acid, 6-chloro, 2-chloro-6-methylbenzoicacid, benzoic acid, 2-chloro-6-methyl, benzoic acid, 6-chloro-4-methyl, 6-chloro-2-methylbenzoic acid, pubchem4568, chloro-6-methylbenzoic acid, 2-m-tolyloxymethyl oxirane | |
| CC1=C(C(=CC=C1)Cl)C(=O)O | |
| 170.592 | |
| 170.59 | |
| Crystalline Powder |
| 21327-86-6 | |
| C8H7ClO2 | |
| 5 g | |
| CEFMMQYDPGCYMG-UHFFFAOYSA-N | |
| 2-chloro-6-methylbenzoic acid | |
| 88870 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 21327-86-6 | |
| 170.592 | |
| CEFMMQYDPGCYMG-UHFFFAOYSA-N | |
| 88870 | |
| CC1=C(C(=CC=C1)Cl)C(=O)O |
| C8H7ClO2 | |
| MFCD00045799 | |
| 6-chloro-o-toluic acid, 2-chloro-6-methyl-benzoic acid, o-toluic acid, 6-chloro, 2-chloro-6-methylbenzoicacid, benzoic acid, 2-chloro-6-methyl, benzoic acid, 6-chloro-4-methyl, 6-chloro-2-methylbenzoic acid, pubchem4568, chloro-6-methylbenzoic acid, 2-m-tolyloxymethyl oxirane | |
| 2-chloro-6-methylbenzoic acid |
Safety and Handling
RTECSNumber : XU1645100
TSCA : No