missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7125648
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27761G 1 g
C27765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27761G Supplier TCI America Supplier No. C27761G

Chemical Identifiers

CAS 21327-86-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00045799
InChI Key CEFMMQYDPGCYMG-UHFFFAOYSA-N
Synonym 6-chloro-o-toluic acid, 2-chloro-6-methyl-benzoic acid, o-toluic acid, 6-chloro, 2-chloro-6-methylbenzoicacid, benzoic acid, 2-chloro-6-methyl, benzoic acid, 6-chloro-4-methyl, 6-chloro-2-methylbenzoic acid, pubchem4568, chloro-6-methylbenzoic acid, 2-m-tolyloxymethyl oxirane
PubChem CID 88870
IUPAC Name 2-chloro-6-methylbenzoic acid
SMILES CC1=C(C(=CC=C1)Cl)C(=O)O

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylbenzoic Acid
RTECSNumber XU1645100
TSCA No