missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzylideneacetone, 98%

Catalog No. AAA1508303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1508303 1 g
AAA1508306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1508303 Supplier Thermo Scientific Chemicals Supplier No. A1508303

CAS: 175136-82-0 | C10H8ClFO | 198.621 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175136-82-0
Molecular Formula C10H8ClFO
Molecular Weight (g/mol) 198.621
MDL Number MFCD00051608
InChI Key VZRBDBXFAKDNDJ-AATRIKPKSA-N
Synonym 2-chloro-6-fluorobenzylideneacetone, 3e-4-2-chloro-6-fluorophenyl but-3-en-2-one, 4-2-chloro-6-fluorophenyl but-3-en-2-one, 1-2-chloro-6-fluorophenyl but-1-en-3-one, e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one, labotest-bb lt00455271, 1-2-chloro-6-fluorophenyl-1-buten-2-one, e-4-2-chloro-6-fluorophenyl-3-buten-2-one, e-4-2-chloro-6-fluorophenyl but-3-en-2-one
PubChem CID 5373895
IUPAC Name (E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=C(C=CC=C1Cl)F

Specifications

Boiling Point 115°C (0.8 mmHg)
Refractive Index 1.58
Quantity 1 g
Formula Weight 198.63
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzylideneacetone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only