CAS RN 175136-82-0
IUPAC Name:
(3E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
Synonyms:
(3E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one
Molecular Weight (g/mol):
198.62
Molecular Formula:
C10H8ClFO
InChi Key:
VZRBDBXFAKDNDJ-AATRIKPKSA-N
SMILES:
CC(=O)\C=C\C1=C(F)C=CC=C1Cl
1
–
1
of
1
results
1
Promotions Available
2-Chloro-6-fluorobenzylideneacetone, 98%
2-Chloro-6-fluorobenzylideneacetone, 98%
CAS: 175136-82-0 Molecular Formula: C10H8ClFO Molecular Weight (g/mol): 198.621 MDL Number: MFCD00051608 InChI Key: VZRBDBXFAKDNDJ-AATRIKPKSA-N Synonym: 2-chloro-6-fluorobenzylideneacetone,3e-4-2-chloro-6-fluorophenyl but-3-en-2-one,4-2-chloro-6-fluorophenyl but-3-en-2-one,1-2-chloro-6-fluorophenyl but-1-en-3-one,e-4-2-chloro-6-fluoro-phenyl but-3-en-2-one,labotest-bb lt00455271,1-2-chloro-6-fluorophenyl-1-buten-2-one,e-4-2-chloro-6-fluorophenyl-3-buten-2-one,e-4-2-chloro-6-fluorophenyl but-3-en-2-one PubChem CID: 5373895 IUPAC Name: (E)-4-(2-chloro-6-fluorophenyl)but-3-en-2-one SMILES: CC(=O)C=CC1=C(C=CC=C1Cl)F