missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluorobenzamide, 98%

Catalog No. AAB2129406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2129406 5 g
AAB2129414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2129406 Supplier Thermo Scientific Chemicals Supplier No. B2129406
Only null left

CAS: 66073-54-9 | C7H5ClFNO | 173.571 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66073-54-9
Molecular Formula C7H5ClFNO
Molecular Weight (g/mol) 173.571
MDL Number MFCD00055436
InChI Key KLOZZZNFJYMTNE-UHFFFAOYSA-N
PubChem CID 98564
IUPAC Name 2-chloro-6-fluorobenzamide
SMILES C1=CC(=C(C(=C1)Cl)C(=O)N)F

Specifications

Melting Point 139°C to 142°C
Quantity 5 g
Synonym 2-fluoro-6-chlorobenzamide, 2-chloro-6-fluoro-benzamide, benzamide, 2-chloro-6-fluoro, acmc-209nth, 6-chloro-2-fluorobenzamide, 2-fluoro-6-chlorobenzoylamino, benzamide,2-chloro-6-fluoro, 2-chloranyl-6-fluoranyl-benzamide
Formula Weight 173.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-fluorobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 266-123-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only