missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluorobenzaldehyde, 95%

Catalog No. AC161510250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161510250 25 g Glass bottle
AC161511000 100 g Glass bottle
AC161515000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC161510250 Supplier Thermo Scientific Chemicals Supplier No. 161510250
Only null left

CAS: 387-45-1 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 387-45-1
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD00003306
InChI Key OACPOWYLLGHGCR-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-6-fluoro, 2,6-cfad, 2-chloro-6-fluoro-benzaldehyd, unii-51yj9bw8w7, 2-chloro-6-fluoro benzaldehyde, 2-chloro-6-fluoro-benzaldehyde, 2-fluoro-6-chloro benzaldehyde, 6-chloro-2-fluorobenzaldehyde, pubchem1434, acmc-209j1r
PubChem CID 67847
IUPAC Name 2-chloro-6-fluorobenzaldehyde
SMILES FC1=CC=CC(Cl)=C1C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 34°C to 39°C
Color White to Yellow
Boiling Point 92°C (10.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CHO
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,ether,chloroform
Formula Weight 158.56
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 2-Chloro-6-fluorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-860-5

RUO – Research Use Only