accessibility menu, dialog, popup

UserName

FClO

CAS RN 387-45-1

IUPAC Name: 2-chloro-6-fluorobenzaldehyde
Synonyms: 2-chloro-6-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: OACPOWYLLGHGCR-UHFFFAOYSA-N
SMILES: FC1=CC=CC(Cl)=C1C=O
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)