missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-(trifluoromethyl)phenol 98.0+%, TCI America™
Supplier: TCI America C294725G
Specifications
| 2-Chloro-5-(trifluoromethyl)phenol | |
| Yellow | |
| C7H4ClF3O | |
| 25 g | |
| 2-chloro-5-trifluoromethyl phenol, 4-chloro-3-hydroxybenzotrifluoride, phenol, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-phenol, 3-hydroxy-4-chlorobenzotrifluoride, m-cresol, 6-chloro-.alpha.,.alpha.,.alpha.-trifluoro-,, pubchem2851, 2-chloranyl-5-trifluoromethyl phenol, 2-chloro-5-trifluoromethyl phenol # | |
| C1=CC(=C(C=C1C(F)(F)F)O)Cl | |
| 196.553 | |
| 196.55 | |
| Liquid |
| 40889-91-6 | |
| 88°C | |
| MFCD00019995 | |
| 1993 | |
| SUIZARUATMVYRT-UHFFFAOYSA-N | |
| 2-chloro-5-(trifluoromethyl)phenol | |
| 123498 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 40889-91-6 | |
| 196.553 | |
| SUIZARUATMVYRT-UHFFFAOYSA-N | |
| 123498 | |
| C1=CC(=C(C=C1C(F)(F)F)O)Cl |
| C7H4ClF3O | |
| MFCD00019995 | |
| 2-chloro-5-trifluoromethyl phenol, 4-chloro-3-hydroxybenzotrifluoride, phenol, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-phenol, 3-hydroxy-4-chlorobenzotrifluoride, m-cresol, 6-chloro-.alpha.,.alpha.,.alpha.-trifluoro-,, pubchem2851, 2-chloranyl-5-trifluoromethyl phenol, 2-chloro-5-trifluoromethyl phenol # | |
| 2-chloro-5-(trifluoromethyl)phenol |
Safety and Handling
EINECSNumber : (3)-0902
TSCA : No