missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)phenol 98.0+%, TCI America™

Catalog No. C29475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29475G 5 g
C294725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29475G Supplier TCI America Supplier No. C29475G

Chemical Identifiers

CAS 40889-91-6
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.553
MDL Number MFCD00019995
InChI Key SUIZARUATMVYRT-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl phenol, 4-chloro-3-hydroxybenzotrifluoride, phenol, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-phenol, 3-hydroxy-4-chlorobenzotrifluoride, m-cresol, 6-chloro-.alpha.,.alpha.,.alpha.-trifluoro-,, pubchem2851, 2-chloranyl-5-trifluoromethyl phenol, 2-chloro-5-trifluoromethyl phenol #
PubChem CID 123498
IUPAC Name 2-chloro-5-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)O)Cl

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
UN Number 1993
Formula Weight 196.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenol
EINECSNumber (3)-0902
TSCA No