Learn More
2-Chloro-5-(trifluoromethyl)aniline, 97%
CAS: 121-50-6 | C7H5ClF3N | 195.569 g/mol
$104.86 - $104.86
Chemical Identifiers
| CAS | 121-50-6 |
|---|---|
| Molecular Formula | C7H5ClF3N |
| Molecular Weight (g/mol) | 195.569 |
| MDL Number | MFCD00007673 |
| InChI Key | VKTTYIXIDXWHKW-UHFFFAOYSA-N |
| Synonym | 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda |
| PubChem CID | 8475 |
| IUPAC Name | 2-chloro-5-(trifluoromethyl)aniline |
| SMILES | C1=CC(=C(C=C1C(F)(F)F)N)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1600422
|
Thermo Scientific Chemicals
A1600422 |
100 g |
Each for $104.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 121-50-6 | |
| 195.569 | |
| VKTTYIXIDXWHKW-UHFFFAOYSA-N | |
| 8475 | |
| C1=CC(=C(C=C1C(F)(F)F)N)Cl |
| C7H5ClF3N | |
| MFCD00007673 | |
| 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda | |
| 2-chloro-5-(trifluoromethyl)aniline |
Specifications
| 121-50-6 | |
| 1.424 | |
| 75°C (167°F) | |
| 1.499 | |
| 100 g | |
| 879910 | |
| VKTTYIXIDXWHKW-UHFFFAOYSA-N | |
| 2-chloro-5-(trifluoromethyl)aniline | |
| 8475 | |
| 97% |
| 8°C to 10°C | |
| 82°C to 83°C (9 mmHg) | |
| C7H5ClF3N | |
| MFCD00007673 | |
| UN2810 | |
| 3-amino-4-chlorobenzotrifluoride, 2-chloro-5-trifluoromethyl aniline, fast orange rd oil, fast orange salt rd, orange salt nrd, benzenamine, 2-chloro-5-trifluoromethyl, fast orange rd salt, daito orange salt rd, diazo fast orange rd, fast orange salt rda | |
| C1=CC(=C(C=C1C(F)(F)F)N)Cl | |
| 195.569 | |
| 195.57 | |
| 2-Chloro-5-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 204-475-7
RTECSNumber : XU5775000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only