missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzophenone 98.0+%, TCI America™

Catalog No. C17765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17765G 5 g
C177625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17765G Supplier TCI America Supplier No. C17765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34052-37-4
Molecular Formula C13H8ClNO3
Molecular Weight (g/mol) 261.661
MDL Number MFCD00007295
InChI Key HRPHZUAPQWJPCZ-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzophenone, 2-chloro-5-nitrophenyl phenyl methanone, 2-chloro-5-nitrophenyl-phenylmethanone, 2-chloro-5-nitrophenyl phenyl ketone, acmc-1aifx, methanone, 2-chloro-5-nitrophenyl phenyl, 5-nitro-2-chlorobenzophenone, 2-chloro-5-nitro-benzophenone, hrphzuapqwjpcz-uhfffaoysa
PubChem CID 118591
IUPAC Name (2-chloro-5-nitrophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)[N+](=O)[O-])Cl

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 261.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzophenone