missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzaldehyde 96.0+%, TCI America™

Catalog No. p-7124288
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C021925G 25 g
C0219250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C021925G Supplier TCI America Supplier No. C021925G

Chemical Identifiers

CAS 6361-21-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007293
InChI Key VFVHWCKUHAEDMY-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
PubChem CID 72933
IUPAC Name 2-chloro-5-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(Cl)C(C=O)=C1

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 25 g
Formula Weight 185.56
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzaldehyde
RTECSNumber CU5220000
TSCA Yes