accessibility menu, dialog, popup

UserName

CAS RN 6361-21-3

IUPAC Name: 2-chloro-5-nitrobenzaldehyde
Synonyms: 2-chloro-5-nitrobenzaldehyde
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: VFVHWCKUHAEDMY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(C=O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)