missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzaldehyde, 97%

Catalog No. p-7022181
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1482409 10 g
AAA1482418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1482409 Supplier Thermo Scientific Chemicals Supplier No. A1482409
Only null left

CAS: 6361-21-3 | C7H4ClNO3 | 185.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6361-21-3
Molecular Formula C7H4ClNO3
Molecular Weight (g/mol) 185.56
MDL Number MFCD00007293
InChI Key VFVHWCKUHAEDMY-UHFFFAOYSA-N
Synonym benzaldehyde, 2-chloro-5-nitro, 3-nitro-6-chlorobenzaldehyde, 2-chloro-5-nitro-benzaldehyde, unii-pf4ill0i3h, pf4ill0i3h, 2-chlor-5-nitrobenzaldehyd, pubchem7375, wln: wnr dg cvh, chloro-5-nitrobenzaldehyde, asischem r37046
PubChem CID 72933
IUPAC Name 2-chloro-5-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(Cl)C(C=O)=C1

Specifications

Melting Point 77°C to 81°C
Quantity 10 g
Beilstein 743764
Sensitivity Air Sensitive
Formula Weight 185.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-nitrobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-830-0
RTECSNumber CU5220000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only