missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylaniline, 98%

Catalog No. p-7028801
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2620906 5 g
AAH2620914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2620906 Supplier Thermo Scientific Chemicals Supplier No. H2620906
Only null left

CAS: 95-81-8 | C7H8ClN | 141.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-81-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007674
InChI Key HPSCXFOQUFPEPE-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-5-methyl, 6-chloro-m-toluidine, 3-amino-4-chlorotoluene, 2-chlor-5-methyl-aniline, 2-chloro-5-methylbenzenamine, 2-chloro-5-methylphenylamine, ccris 2888, 2-chloro-5-methyl aniline, 6-chlor-meta-toluidin, pubchem18921
PubChem CID 66770
IUPAC Name 2-chloro-5-methylaniline
SMILES CC1=CC=C(Cl)C(N)=C1

Specifications

Melting Point 31°C to 33°C
Boiling Point 228°C to 230°C
Flash Point 107°C (225°F)
Quantity 5 g
UN Number UN2239
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID
EINECSNumber 202-454-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only