missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methoxyphenol 97.0+%, TCI America™
$123.21 - $123.21
Chemical Identifiers
| CAS | 18113-04-7 |
|---|---|
| Molecular Formula | C7H7ClO2 |
| Molecular Weight (g/mol) | 158.581 |
| MDL Number | MFCD00086345 |
| InChI Key | KULSVCANYQIOFD-UHFFFAOYSA-N |
| Synonym | 4-Chloro-3-hydroxyanisole |
| PubChem CID | 140336 |
| IUPAC Name | 2-chloro-5-methoxyphenol |
| SMILES | COC1=CC(=C(C=C1)Cl)O |
Chemical Identifiers
| 18113-04-7 | |
| 158.581 | |
| KULSVCANYQIOFD-UHFFFAOYSA-N | |
| 140336 | |
| COC1=CC(=C(C=C1)Cl)O |
| C7H7ClO2 | |
| MFCD00086345 | |
| 4-Chloro-3-hydroxyanisole | |
| 2-chloro-5-methoxyphenol |
Specifications
| 18113-04-7 | |
| 125°C | |
| MFCD00086345 | |
| 4-Chloro-3-hydroxyanisole | |
| COC1=CC(=C(C=C1)Cl)O | |
| 158.581 | |
| 158.58 | |
| Liquid |
| Yellow | |
| C7H7ClO2 | |
| 5 g | |
| KULSVCANYQIOFD-UHFFFAOYSA-N | |
| 2-chloro-5-methoxyphenol | |
| 140336 | |
| ≥97.0% (GC) | |
| 2-Chloro-5-methoxyphenol |
Safety and Handling
TSCA : No