Learn More
2-Chloro-5-methoxyaniline, 98+%
CAS: 2401-24-3 | C7H8ClNO | 157.597 g/mol
Supplier: Thermo Scientific Chemicals L2016214
Description
2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| 2-Chloro-5-methoxyaniline | |
| 27°C | |
| Characteristic | |
| 1.5865 | |
| 25 g | |
| 2082193 | |
| Slightly soluble in water. | |
| COC1=CC(=C(C=C1)Cl)N | |
| 157.597 | |
| 157.6 |
| 2401-24-3 | |
| 135°C to 137°C (12 mmHg) | |
| C7H8ClNO | |
| MFCD00047830 | |
| UN2233 | |
| 6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038 | |
| GBOUQGUQUUPGLO-UHFFFAOYSA-N | |
| 2-chloro-5-methoxyaniline | |
| 75460 | |
| ≥98% |
Chemical Identifiers
| 2401-24-3 | |
| 157.597 | |
| GBOUQGUQUUPGLO-UHFFFAOYSA-N | |
| 75460 | |
| COC1=CC(=C(C=C1)Cl)N |
| C7H8ClNO | |
| MFCD00047830 | |
| 6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038 | |
| 2-chloro-5-methoxyaniline |
Safety and Handling
GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROANISIDINES
EINECSNumber : 219-277-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only