missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxyaniline, 98+%

Catalog No. p-7048053
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2016206 5 g
AAL2016214 25 g
AAL2016222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2016206 Supplier Thermo Scientific Chemicals Supplier No. L2016206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2401-24-3 | C7H8ClNO | 157.597 g/mol

2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-methoxyaniline is used in the preparation of 4-amino-5-chloro-2-methoxyphenyl thiocyanate.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2401-24-3
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00047830
InChI Key GBOUQGUQUUPGLO-UHFFFAOYSA-N
Synonym 6-chloro-m-anisidine, 3-amino-4-chloroanisole, benzenamine, 2-chloro-5-methoxy, 2-chloro-5-methoxy-aniline, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxy-phenylamine, 2-chloro-5-methoxybenzenamine hydrochloride, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, timtec-bb sbb008038
PubChem CID 75460
IUPAC Name 2-chloro-5-methoxyaniline
SMILES COC1=CC(=C(C=C1)Cl)N

Specifications

Melting Point 27°C
Boiling Point 135°C to 137°C (12 mmHg)
Odor Characteristic
Refractive Index 1.5865
Quantity 5 g
UN Number UN2233
Beilstein 2082193
Solubility Information Slightly soluble in water.
Formula Weight 157.6
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-5-methoxyaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROANISIDINES
EINECSNumber 219-277-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only