missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodobenzoic acid, 97%

Catalog No. p-7048052
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2016106 5 g
AAL2016114 25 g
AAL2016122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2016106 Supplier Thermo Scientific Chemicals Supplier No. L2016106
Only null left

CAS: 19094-56-5 | C7H4ClIO2 | 282.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19094-56-5
Molecular Formula C7H4ClIO2
Molecular Weight (g/mol) 282.46
MDL Number MFCD00079731
InChI Key GEBYSTBEDVQOTK-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-benzoic acid, benzoic acid, 2-chloro-5-iodo, pubchem3649, acmc-209eug, 5-iodo-2-chlorobenzoic acid, ksc174i1h, benzoicacid, 2-chloro-5-iodo, buttpark 100\01-43, 2-chloro-5-iodobenzoic acid
PubChem CID 519638
IUPAC Name 2-chloro-5-iodobenzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(I)=C1

Specifications

Melting Point 156°C to 161°C
Quantity 5 g
Beilstein 2254110
Sensitivity Light sensitive
Formula Weight 282.46
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only