missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxybenzotrifluoride 98.0+%, TCI America™

Catalog No. C15165G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15165G 5 g
C15161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15165G Supplier TCI America Supplier No. C15165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6294-93-5
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.55
MDL Number MFCD00042525
InChI Key ZLFPIEUWXNRPNM-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenol, 2-chloro-5-hydroxybenzotrifluoride, phenol, 4-chloro-3-trifluoromethyl, 4-chloro-3-trifluoromethyl-phenol, phenol, p-chloro-m-trifluoromethyl, p-chloro-m-trifluoromethylphenol, 2-chloro-5-hydroxybezotrifluoride, pubchem2863
PubChem CID 80520
IUPAC Name 4-chloro-3-(trifluoromethyl)phenol
SMILES OC1=CC=C(Cl)C(=C1)C(F)(F)F

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 122°C
Quantity 5 g
Formula Weight 196.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-hydroxybenzotrifluoride
RTECSNumber SK6040000
TSCA No