CAS RN 6294-93-5
IUPAC Name:
4-chloro-3-(trifluoromethyl)phenol
Synonyms:
4-chloro-3-(trifluoromethyl)phenol
Molecular Weight (g/mol):
196.55
Molecular Formula:
C7H4ClF3O
InChi Key:
ZLFPIEUWXNRPNM-UHFFFAOYSA-N
SMILES:
OC1=CC=C(Cl)C(=C1)C(F)(F)F
1
–
1
of
1
results
1
2-Chloro-5-hydroxybenzotrifluoride 98.0+%, TCI America™
CAS: 6294-93-5 Molecular Formula: C7H4ClF3O Molecular Weight (g/mol): 196.55 MDL Number: MFCD00042525 InChI Key: ZLFPIEUWXNRPNM-UHFFFAOYSA-N Synonym: 4-chloro-3-trifluoromethyl phenol,2-chloro-5-hydroxybenzotrifluoride,phenol, 4-chloro-3-trifluoromethyl,4-chloro-3-trifluoromethyl-phenol,phenol, p-chloro-m-trifluoromethyl,p-chloro-m-trifluoromethylphenol,2-chloro-5-hydroxybezotrifluoride,pubchem2863 PubChem CID: 80520 IUPAC Name: 4-chloro-3-(trifluoromethyl)phenol SMILES: OC1=CC=C(Cl)C(=C1)C(F)(F)F