missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-ethylpyrimidine 98.0+%, TCI America™
$46.60 - $130.48
Chemical Identifiers
| CAS | 111196-81-7 |
|---|---|
| Molecular Formula | C6H7ClN2 |
| Molecular Weight (g/mol) | 142.59 |
| MDL Number | MFCD00799503 |
| InChI Key | BGLLZQRUXJGTAD-UHFFFAOYSA-N |
| Synonym | 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b |
| PubChem CID | 3572763 |
| IUPAC Name | 2-chloro-5-ethylpyrimidine |
| SMILES | CCC1=CN=C(Cl)N=C1 |
Chemical Identifiers
| 111196-81-7 | |
| 142.59 | |
| BGLLZQRUXJGTAD-UHFFFAOYSA-N | |
| 3572763 | |
| CCC1=CN=C(Cl)N=C1 |
| C6H7ClN2 | |
| MFCD00799503 | |
| 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b | |
| 2-chloro-5-ethylpyrimidine |
Specifications
| 111196-81-7 | |
| 86°C | |
| MFCD00799503 | |
| 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b | |
| CCC1=CN=C(Cl)N=C1 | |
| 142.59 | |
| 142.59 | |
| Liquid |
| Yellow | |
| C6H7ClN2 | |
| 1 g | |
| BGLLZQRUXJGTAD-UHFFFAOYSA-N | |
| 2-chloro-5-ethylpyrimidine | |
| 3572763 | |
| ≥98.0% (GC) | |
| 2-Chloro-5-ethylpyrimidine |
Safety and Handling
TSCA : No