accessibility menu, dialog, popup

UserName

H3CNNCl

CAS RN 111196-81-7

IUPAC Name: 2-chloro-5-ethylpyrimidine
Synonyms: 2-chloro-5-ethylpyrimidine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: BGLLZQRUXJGTAD-UHFFFAOYSA-N
SMILES: CCC1=CN=C(Cl)N=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)