missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-ethylpyrimidine 98.0+%, TCI America™

Catalog No. c17545g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17545G 5 g
C17541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17545G Supplier TCI America Supplier No. C17545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111196-81-7
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00799503
InChI Key BGLLZQRUXJGTAD-UHFFFAOYSA-N
Synonym 5-ethyl-2-chloropyrimidine, 2-chloro-5-ethyl-pyrimidine, pyrimidine, 2-chloro-5-ethyl, 2-chloro-5-ethyl pyrimidine, pubchem7032, acmc-1brw8, 2-chloro-5-ethylpyrimdine, 2-chloro-5-ethylpyrimidin, 2-chloro-5-ethylpyrimidine, ksc493s1b
PubChem CID 3572763
IUPAC Name 2-chloro-5-ethylpyrimidine
SMILES CCC1=CN=C(Cl)N=C1

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
Formula Weight 142.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-ethylpyrimidine