missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)phenol 98.0+%, TCI America™

Catalog No. p-7125936
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33555G 5 g
C335525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33555G Supplier TCI America Supplier No. C33555G

Chemical Identifiers

CAS 35852-58-5
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.553
MDL Number MFCD00042217
InChI Key YNWKEXMSQQUMEL-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl phenol, 3-chloro-4-hydroxybenzotrifluoride, 2-chloro-4-trifuloromethyl phenol, phenol, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl-phenol, 2-chloro-4-trifluoromethylphenl, pubchem4105, 2chloro-4-trifluoromethylphenol, chloro-4-trifluoromethyl-phenol
PubChem CID 2736603
IUPAC Name 2-chloro-4-(trifluoromethyl)phenol
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)O

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 g
Formula Weight 196.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)phenol