missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%

CAS: 254993-59-4 | C7H5BClF3O2 | 224.37 g/mol

$88.26 - $294.81

Chemical Identifiers

CAS 254993-59-4
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.37
MDL Number MFCD02684337
InChI Key JKSUCAFAUNSPLO-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl phenylboronic acid, 2-chloro-4-trifluoromethylphenylboronic acid, 2-chloro-4-trifluoromethyl benzeneboronic acid, 2-chloro-4-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl phenylboronicacid, 2-chloro-4-trifluoromethylphenyl boronic acid, boronic acid, 2-chloro-4-trifluoromethyl phenyl, 2-fluoro-4-iodophenylboronicacid
PubChem CID 2783251
IUPAC Name [2-chloro-4-(trifluoromethyl)phenyl]boronic acid
SMILES OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2010303
View Documents
Thermo Scientific Chemicals
L2010303
1 g
Each for $88.26
Only null left
 
AAL2010306
View Documents
Thermo Scientific Chemicals
L2010306
5 g
Each for $294.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

254993-59-4
224.37
JKSUCAFAUNSPLO-UHFFFAOYSA-N
2783251
OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F
C7H5BClF3O2
MFCD02684337
2-chloro-4-trifluoromethyl phenylboronic acid, 2-chloro-4-trifluoromethylphenylboronic acid, 2-chloro-4-trifluoromethyl benzeneboronic acid, 2-chloro-4-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl phenylboronicacid, 2-chloro-4-trifluoromethylphenyl boronic acid, boronic acid, 2-chloro-4-trifluoromethyl phenyl, 2-fluoro-4-iodophenylboronicacid
[2-chloro-4-(trifluoromethyl)phenyl]boronic acid

Specifications

254993-59-4
C7H5BClF3O2
1 g
2-chloro-4-trifluoromethyl phenylboronic acid, 2-chloro-4-trifluoromethylphenylboronic acid, 2-chloro-4-trifluoromethyl benzeneboronic acid, 2-chloro-4-trifluoromethyl phenyl boronic acid, 2-chloro-4-trifluoromethyl phenylboronicacid, 2-chloro-4-trifluoromethylphenyl boronic acid, boronic acid, 2-chloro-4-trifluoromethyl phenyl, 2-fluoro-4-iodophenylboronicacid
OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F
224.37
224.38
2-Chloro-4-(trifluoromethyl)benzeneboronic acid
110°C to 112°C
MFCD02684337
9263388
JKSUCAFAUNSPLO-UHFFFAOYSA-N
[2-chloro-4-(trifluoromethyl)phenyl]boronic acid
2783251
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only