CAS RN 254993-59-4
IUPAC Name:
[2-chloro-4-(trifluoromethyl)phenyl]boronic acid
Synonyms:
2-chloro-4-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
224.37
Molecular Formula:
C7H5BClF3O2
InChi Key:
JKSUCAFAUNSPLO-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%
2-Chloro-4-(trifluoromethyl)benzeneboronic acid, 96%
CAS: 254993-59-4 Molecular Formula: C7H5BClF3O2 Molecular Weight (g/mol): 224.37 MDL Number: MFCD02684337 InChI Key: JKSUCAFAUNSPLO-UHFFFAOYSA-N Synonym: 2-chloro-4-trifluoromethyl phenylboronic acid,2-chloro-4-trifluoromethylphenylboronic acid,2-chloro-4-trifluoromethyl benzeneboronic acid,2-chloro-4-trifluoromethyl phenyl boronic acid,2-chloro-4-trifluoromethyl phenylboronicacid,2-chloro-4-trifluoromethylphenyl boronic acid,boronic acid, 2-chloro-4-trifluoromethyl phenyl,2-fluoro-4-iodophenylboronicacid PubChem CID: 2783251 IUPAC Name: [2-chloro-4-(trifluoromethyl)phenyl]boronic acid SMILES: OB(O)C1=C(Cl)C=C(C=C1)C(F)(F)F